KEY465194019 Key Organics / BIONET

2-sulfanylidene-3-[4-(trifluoromethyl)phenyl]imidazolidin-4-one

  • MDL number MFCD00975555
  • Cas Number : 338773-10-7
  • Molecular Weight : 260.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67739

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2K-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart