KEY465194022 Key Organics / BIONET

N',2-dimethyl-N'-[3-(trifluoromethyl)quinoxalin-2-yl]benzohydrazide

  • MDL number MFCD00974655
  • Cas Number : 338773-26-5
  • Molecular Weight : 360.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67757

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2K-029_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart