KEY465194026 Key Organics / BIONET

4-(dimethylamino)-2-{[4-(trifluoromethyl)phenyl]amino}pyridine-3-carbonitrile

  • MDL number MFCD01314683
  • Cas Number : 338773-40-3
  • Molecular Weight : 306.292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2K-048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart