KEY465194031 Key Organics / BIONET

2,4-bis(trifluoromethyl)-[1,2,4]triazolo[4,3-a]1,8-naphthyridin-9-amine

  • MDL number MFCD00974664
  • Cas Number : 338773-55-0
  • Molecular Weight : 321.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67791

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2K-065_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart