KEY465194046 Key Organics / BIONET

11-(dimethylamino)-6-fluoro-1,8-diazatricyclo[7.4.0.0^{2,7}]trideca-2(7),3,5,8,10,12-hexaene-10-carbonitrile

  • MDL number MFCD01314705
  • Cas Number : 338773-88-9
  • Molecular Weight : 254.268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67824

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2K-098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart