KEY465194047 Key Organics / BIONET

2-[(2E)-3-[(2,4-difluorophenyl)amino]-1-(dimethylamino)prop-2-en-1-ylidene]propanedinitrile

  • MDL number MFCD01314707
  • Cas Number : 1164476-37-2
  • Molecular Weight : 274.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67826

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2K-100_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart