KEY465194049 Key Organics / BIONET

2-[(2E)-3-[(2-acetylphenyl)amino]-1-(dimethylamino)prop-2-en-1-ylidene]propanedinitrile

  • MDL number MFCD01314710
  • Cas Number : 338773-94-7
  • Molecular Weight : 280.331

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67830

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2K-105_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart