KEY465194055 Key Organics / BIONET

6-amino-5-(benzenesulfonyl)-2-methoxypyridine-3-carbonitrile

  • MDL number MFCD00244118
  • Cas Number : 119672-31-0
  • Molecular Weight : 289.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67886

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2K-512S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart