KEY465194056 Key Organics / BIONET

6-amino-2-methoxy-5-[3-(trifluoromethyl)benzenesulfonyl]pyridine-3-carbonitrile

  • MDL number MFCD00244122
  • Cas Number : 338774-45-1
  • Molecular Weight : 357.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67890

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2K-517S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart