KEY465194063 Key Organics / BIONET

N-(2,3-dichlorophenyl)-6-hydroxy-1-methyl-2-oxo-1,2-dihydropyrimidine-4-carboxamide

  • MDL number MFCD00244522
  • Cas Number : 338774-73-5
  • Molecular Weight : 314.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67926

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2K-605S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart