KEY465194064 Key Organics / BIONET

N-(4-bromophenyl)-6-hydroxy-1-methyl-2-oxo-1,2-dihydropyrimidine-4-carboxamide

  • MDL number MFCD00244527
  • Cas Number : 338774-79-1
  • Molecular Weight : 324.134

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67937

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2K-623S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart