KEY465194071 Key Organics / BIONET

3-{5H,6H-benzo[h]quinazolin-2-yl}-1-[3-(trifluoromethyl)phenyl]urea

  • MDL number MFCD00231357
  • Cas Number : 338774-92-8
  • Molecular Weight : 384.362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67954

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2K-900_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart