KEY465194075 Key Organics / BIONET

(2E)-2-(5-amino-1,3,4-thiadiazol-2-yl)-3-(2,4-dichlorophenyl)prop-2-enenitrile

  • MDL number MFCD00663678
  • Cas Number : 338775-21-6
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67989

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2L-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart