KEY465194080 Key Organics / BIONET

(E)-N'-{5-[(1E)-1-cyano-2-(4-methoxyphenyl)eth-1-en-1-yl]-1,3,4-thiadiazol-2-yl}-N,N-dimethylmethanimidamide

  • MDL number MFCD00663682
  • Cas Number : 338775-59-0
  • Molecular Weight : 313.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68015

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2L-047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart