KEY465194084 Key Organics / BIONET

2,4-dichloro-N-[(2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]benzamide

  • MDL number MFCD00794295
  • Cas Number : 477885-66-8
  • Molecular Weight : 344.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68022

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2L-305S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart