KEY465194112 Key Organics / BIONET

N-[(2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]-4-(trifluoromethyl)benzamide

  • MDL number MFCD00794311
  • Cas Number : 477885-90-8
  • Molecular Weight : 343.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68082

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2L-373S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart