KEY465194123 Key Organics / BIONET

3-(4-bromophenyl)-4-(4-chlorobenzoyl)-1H-pyrrole

  • MDL number MFCD00794452
  • Cas Number : 477886-29-6
  • Molecular Weight : 360.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68145

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2L-581S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart