KEY465194125 Key Organics / BIONET

(2E)-3-(4-bromophenyl)-1-(4-chlorophenyl)prop-2-en-1-one

  • MDL number MFCD00018702
  • Cas Number : 126443-20-7
  • Molecular Weight : 321.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68147

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2L-583S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart