KEY465194128 Key Organics / BIONET

(E)-N-hydroxy-N-methyl-N'-(4-methyl-6-oxo-1,6-dihydropyrimidin-2-yl)methanimidamide

  • MDL number MFCD01315933
  • Cas Number : 338400-04-7
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68169

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2L-918_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart