KEY465194131 Key Organics / BIONET

4-hydroxy-2-[4-(propan-2-yl)phenyl]-6H-1,3-thiazin-6-one

  • MDL number MFCD00793779
  • Cas Number : 338400-11-6
  • Molecular Weight : 247.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68178

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2M-006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart