KEY465194132 Key Organics / BIONET

1,3-dimethyl 2-(4-chlorophenoxy)-2-[(3-methylphenyl)methyl]propanedioate

  • MDL number MFCD00793780
  • Cas Number : 338400-12-7
  • Molecular Weight : 362.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68179

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2M-008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart