KEY465194138 Key Organics / BIONET

(2Z)-2-[2-(3-methylphenyl)hydrazin-1-ylidene]-3-oxo-3-(thiophen-2-yl)propanal

  • MDL number MFCD00975085
  • Cas Number : 338400-27-4
  • Molecular Weight : 272.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68197

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2M-032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart