KEY465194139 Key Organics / BIONET

(2Z)-3-(furan-2-yl)-3-oxo-2-(2-phenylhydrazin-1-ylidene)propanal

  • MDL number MFCD00975086
  • Cas Number : 186965-61-7
  • Molecular Weight : 242.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68198

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2M-033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart