KEY465194143 Key Organics / BIONET

(2E,3E)-2-[2-(3,5-dimethylphenyl)hydrazin-1-ylidene]-3-hydrazinylidene-1-(4-methoxyphenyl)propan-1-one

  • MDL number MFCD00975090
  • Cas Number : 338400-36-5
  • Molecular Weight : 324.384

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68207

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2M-044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart