KEY465194144 Key Organics / BIONET

(2E)-2-[2-(3,5-dichlorophenyl)hydrazin-1-ylidene]-3-(4-methylphenyl)-3-oxopropanal

  • MDL number MFCD00975091
  • Cas Number : 338400-38-7
  • Molecular Weight : 335.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68209

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2M-046_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart