KEY465194160 Key Organics / BIONET

1-(4-chlorophenyl)-2-[3-(trifluoromethyl)phenoxy]ethan-1-one

  • MDL number MFCD01314927
  • Cas Number : 338400-72-9
  • Molecular Weight : 314.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68286

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2M-570S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart