KEY465194165 Key Organics / BIONET

4-(3-chloro-1-benzothiophen-2-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarbonitrile

  • MDL number MFCD00794167
  • Cas Number : 477886-59-2
  • Molecular Weight : 325.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68313

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2M-704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart