KEY465194188 Key Organics / BIONET

(2E)-3-(4-chlorophenyl)-1-(3-methoxyphenyl)prop-2-en-1-one

  • MDL number MFCD01315393
  • Cas Number : 130820-52-9
  • Molecular Weight : 272.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68400

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2N-034_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart