KEY465194191 Key Organics / BIONET

(2E)-3-(2-chloro-6-fluorophenyl)-1-(4-fluorophenyl)prop-2-en-1-one

  • MDL number MFCD01315395
  • Cas Number : 1164538-77-5
  • Molecular Weight : 278.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2N-043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart