KEY465194208 Key Organics / BIONET

3-chloro-2-{[4-(prop-2-yn-1-yloxy)phenyl]methyl}-5-(trifluoromethyl)pyridine

  • MDL number MFCD01316647
  • Cas Number : 338402-02-1
  • Molecular Weight : 325.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68509

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2N-734_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart