KEY465194209 Key Organics / BIONET

(2E)-1-(2,5-dichlorothiophen-3-yl)-3-[(3-nitrophenyl)amino]prop-2-en-1-one

  • MDL number MFCD01316649
  • Cas Number : 338402-04-3
  • Molecular Weight : 343.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68511

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2N-736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart