KEY465194212 Key Organics / BIONET

N-cyclohexyl-2-(trifluoromethyl)pyridin-4-amine

  • MDL number MFCD01568337
  • Cas Number : 338402-06-5
  • Molecular Weight : 244.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68514

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2N-739_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart