KEY465194222 Key Organics / BIONET

[(Z)-(1-{2-[(2-chloro-1,3-thiazol-5-yl)methoxy]phenyl}ethylidene)amino]thiourea

  • MDL number MFCD01814632
  • Cas Number : 477886-88-7
  • Molecular Weight : 340.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68579

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-038_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart