KEY465194243 Key Organics / BIONET

N'-[(1E)-(3-phenyl-1H-pyrazol-4-yl)methylidene]-4-(trifluoromethyl)benzohydrazide

  • MDL number MFCD01814791
  • Cas Number : 477710-90-0
  • Molecular Weight : 358.324

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68625

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-093_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart