KEY465194251 Key Organics / BIONET

methyl 2-[2-(dimethylsulfamoyl)-4,5-dimethoxyphenyl]acetate

  • MDL number MFCD02082172
  • Cas Number : 477887-67-5
  • Molecular Weight : 317.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68692

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-519S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart