KEY465194256 Key Organics / BIONET

methyl 3-{2-[4-(4-fluorophenyl)piperazin-1-yl]acetamido}thiophene-2-carboxylate

  • MDL number MFCD01814746
  • Cas Number : 227958-59-0
  • Molecular Weight : 377.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68699

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-530S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart