KEY465194257 Key Organics / BIONET

methyl 3-{2-[4-(3-chlorophenyl)piperazin-1-yl]acetamido}thiophene-2-carboxylate

  • MDL number MFCD01833279
  • Cas Number : 477887-73-3
  • Molecular Weight : 393.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68701

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-532S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart