KEY465194259 Key Organics / BIONET

1-(4-fluorophenyl)-2-phenyl-4,5,6,7-tetrahydro-1H-indol-4-one

  • MDL number MFCD01815013
  • Cas Number : 96757-16-3
  • Molecular Weight : 305.352

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-572S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart