KEY465194261 Key Organics / BIONET

1,3-dimethyl-4-[(E)-2-nitroethenyl]-5-phenoxy-1H-pyrazole

  • MDL number MFCD01814748
  • Cas Number : 477710-91-1
  • Molecular Weight : 259.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68714

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-582S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart