KEY465194262 Key Organics / BIONET

5-{4H,5H-naphtho[1,2-b]thiophen-2-yl}-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD01815423
  • Cas Number : 477887-99-3
  • Molecular Weight : 286.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68735

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-667S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart