KEY465194265 Key Organics / BIONET

ethyl 3-(2-chloro-6-fluorophenyl)-2-(1,2-oxazol-5-yl)propanoate

  • MDL number MFCD02082393
  • Cas Number : 477888-06-5
  • Molecular Weight : 297.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68743

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-707_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart