KEY465194266 Key Organics / BIONET

(2E)-1-(4-fluorophenyl)-3-[(2-hydroxyphenyl)amino]prop-2-en-1-one

  • MDL number MFCD01814792
  • Cas Number : 423747-09-5
  • Molecular Weight : 257.264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68747

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart