KEY465194269 Key Organics / BIONET

ethyl 2-[1-(3-nitrophenyl)-1H-pyrazol-5-yl]-3-phenylpropanoate

  • MDL number MFCD01814798
  • Cas Number : 477710-93-3
  • Molecular Weight : 365.389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68762

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart