KEY465194271 Key Organics / BIONET

1-[(3,4-dimethoxyphenyl)methyl]-4-hydroxy-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one

  • MDL number MFCD02082399
  • Cas Number : 477888-18-9
  • Molecular Weight : 377.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68766

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-741_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart