KEY465194277 Key Organics / BIONET

ethyl 2-[(2-benzoyl-3-oxo-3-phenylprop-1-en-1-yl)amino]acetate

  • MDL number MFCD01814803
  • Cas Number : 220957-52-8
  • Molecular Weight : 337.375

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-804_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart