KEY465194278 Key Organics / BIONET

N-(prop-2-en-1-yl)-N'-(propan-2-ylidene)formohydrazide

  • MDL number MFCD01814635
  • Cas Number : 141499-25-4
  • Molecular Weight : 140.186

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68775

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-805_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart