KEY465194281 Key Organics / BIONET

2-[4-(4-chlorophenyl)-2-methyl-3-oxo-1,2,4-oxadiazolidin-5-yl]-N-(4-methoxyphenyl)acetamide

  • MDL number MFCD02082402
  • Cas Number : 477888-23-6
  • Molecular Weight : 375.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68780

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-811_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart