KEY465194292 Key Organics / BIONET

(2Z)-2-[4-(3-fluoropropoxy)phenyl]-3-{[4-(trifluoromethoxy)phenyl]amino}prop-2-enenitrile

  • MDL number MFCD02102542
  • Cas Number : 477888-58-7
  • Molecular Weight : 380.343

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68833

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-0059_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart