KEY465194298 Key Organics / BIONET

(1E)-1-[(4-chlorophenyl)amino]-4-({2-fluoro-[1,1'-biphenyl]-4-yl}oxy)pent-1-en-3-one

  • MDL number MFCD02102562
  • Cas Number : 477888-81-6
  • Molecular Weight : 395.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68862

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0096_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart