KEY465194312 Key Organics / BIONET

(1R,6S)-6-[(2-{[(1S,6R)-6-carboxycyclohex-3-en-1-yl]formamido}ethyl)carbamoyl]cyclohex-3-ene-1-carboxylic acid

  • MDL number MFCD08690893
  • Cas Number : 415973-33-0
  • Molecular Weight : 364.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68910

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0164_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart